Please login to your online account to display your discounted pricing

Potassium Phosphate Monobasic, ≥98%, MP Biomedicals


Quantity 1kg

Manufacturer: mp biomedicals  0219484691

 View more versions of this product

Catalog No. ICN19484691

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1kg
Chemical Name or Material Potassium Phosphate Monobasic
Name Note Anhydrous
CAS 7778-77-0
Assay Percent Range ≥98%
Molecular Formula KH2PO4
Formula Weight 136.09g/mol
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Potassium Phosphate Monobasic
Name Note Anhydrous
CAS 7778-77-0
Assay Percent Range ≥98%
Molecular Formula KH2PO4
Formula Weight 136.09g/mol
Accessories

Room Temperature