Please login to your online account to display your discounted pricing

Potassium Phosphate Monobasic, ≥98%, MP Biomedicals


Quantity 500g

Manufacturer: mp biomedicals  19484690

 View more versions of this product

Catalog No. ICN19484690

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 136.09
Quantity 500g
Assay Percent Range ≥98%
CAS 7778-77-0
Recommended Storage Room Temperature
Name Note Anhydrous
Molecular Formula KH2PO4
Chemical Name or Material Potassium Phosphate Monobasic

Specifications

Chemical Name or Material Potassium Phosphate Monobasic
Name Note Anhydrous
CAS 7778-77-0
Assay Percent Range ≥98%
Molecular Formula KH2PO4
Formula Weight 136.09
Accessories

Room Temperature