Please login to your online account to display your discounted pricing

Potassium Phosphate Monobasic, ≥98%, MP Biomedicals

$109.44 - $109.44

Chemical Identifiers

Chemical Name or Material Potassium Phosphate Monobasic
Name Note Anhydrous
CAS 7778-77-0
Assay Percent Range ≥98%
Molecular Formula KH2PO4
Formula Weight 136.09
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19484690

 
mp biomedicals
19484690
500g Each for $109.44
Description & Specifications

Specifications

Formula Weight 136.09
Quantity 500g
Assay Percent Range ≥98%
CAS 7778-77-0
Recommended Storage Room Temperature
Name Note Anhydrous
Molecular Formula KH2PO4
Chemical Name or Material Potassium Phosphate Monobasic