missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals Inc Potassium Phosphate Dibasic Trihydrate, 99+%, (Crystalline), MP Biomedicals
Supplier: MP Biomedicals Inc 0215194610
Specifications
Potassium Phosphate Dibasic Trihydrate | |
Crystalline | |
99+% | |
MFCD00149926 | |
XQGPKZUNMMFTAL-UHFFFAOYSA-L | |
dipotassium trihydrate hydrogen phosphate | |
16217523 |
16788-57-1 | |
10 kg | |
H7K2O7P | |
dipotassium hydrogen phosphate trihydrate, dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate | |
O.O.O.[K+].[K+].OP([O-])([O-])=O | |
228.22 | |
228.2 |
Chemical Identifiers
16788-57-1 | |
228.22 | |
XQGPKZUNMMFTAL-UHFFFAOYSA-L | |
16217523 | |
O.O.O.[K+].[K+].OP([O-])([O-])=O |
H7K2O7P | |
MFCD00149926 | |
dipotassium hydrogen phosphate trihydrate, dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate | |
dipotassium trihydrate hydrogen phosphate |
Safety and Handling
Recommended Storage : Room Temperature