missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium phosphate, dibasic trihydrate, 99+%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 205930025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferSpecifications
Potassium phosphate, dibasic trihydrate | |
White | |
8.5 to 9.6 (5% soln.) | |
2.5 kg | |
H7K2O7P | |
MFCD00149926 | |
dipotassium hydrogen phosphate trihydrate, dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate | |
XQGPKZUNMMFTAL-UHFFFAOYSA-L | |
dipotassium;hydrogen phosphate;trihydrate | |
16217523 | |
≥99% | |
Plastic bottle |
99+% | |
16788-57-1 | |
Adhering Crystals | |
99+% | |
K2HPO4·3H2O | |
10, 7541 | |
Solubility in water: freely soluble | |
O.O.O.[K+].[K+].OP([O-])([O-])=O | |
228.22 | |
228.23 | |
Analytical |
Chemical Identifiers
16788-57-1 | |
228.22 | |
XQGPKZUNMMFTAL-UHFFFAOYSA-L | |
16217523 |
H7K2O7P | |
MFCD00149926 | |
dipotassium hydrogen phosphate trihydrate, dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate | |
O.O.O.[K+].[K+].OP([O-])([O-])=O |
RUO – Research Use Only