missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Phosphate Dibasic Anhydrous (Crystalline Powder/Certified ACS), Fisher Chemical

Manufacturer:  Thermo Scientific FLP288200LB

 View more versions of this product

Catalog No. P288-200LB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Potassium Phosphate Dibasic Anhydrous
Certified ACS Grade
≥98.0 %
MFCD00011383
Soluble in water
OP(=O)([O-])[O-].[K+].[K+]
174.17
CHEBI:32031
Solid
5ppm max.
0.01% max.
1% max.
Fiber Drum
0.005% max.
White
8.5 to 9.6
7758-11-4
≥98.0%
HK2O4P
Potassium Monohydrogen Phosphate, Dipotassium Hydrogen Phosphate
ZPWVASYFFYYZEW-UHFFFAOYSA-L
dipotassium;hydrogen phosphate
24450
174.18
0.003% max.
Pass Test
0.001% max.
0.001% max.
0.05% max.
>180°C
340°C
200 lb.

Chemical Identifiers

7758-11-4
174.17
ZPWVASYFFYYZEW-UHFFFAOYSA-L
24450
dipotassium;hydrogen phosphate
HK2O4P
MFCD00011383
Potassium Monohydrogen Phosphate, Dipotassium Hydrogen Phosphate
CHEBI:32031
OP(=O)([O-])[O-].[K+].[K+]
Safety and Handling

Safety and Handling

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:1
Instability:0

CAUTION!

SDS Documents