missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Phosphate, Dibasic, ACS Reagent Grade, Ricca Chemical

Chemical Identifiers

CAS 7758-11-4
CAS Min % 98.0
CAS Max % 100.0
Physical Form Powder or Cyrstals
Chemical Name or Material Potassium Phosphate, Dibasic
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
RDCP0650500 Ricca Chemical Company
RIRDCP0650500B1
Poly wht WM 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
RDCP065050F Ricca Chemical Company
RIRDCP065050F4
Drum, Fiber 50kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7758-11-4
100.0
Potassium Phosphate, Dibasic
98.0
Powder or Cyrstals
ACS Grade

Specifications

500g
White
100.0
98.0
ACS Grade
Powder or Cyrstals
7758-11-4
Poly wht WM
Potassium Phosphate, Dibasic

Safety and Handling

ShelfLife : 60 months