Please login to your online account to display your discounted pricing

Potassium Phosphate, Dibasic, ACS Grade, ≥ 98.0%, LabChem™

$172.63 - $172.63

Chemical Identifiers

Chemical Name or Material Potassium Phosphate
CAS 7758-11-4
CAS Max % 100
Name Note Dibasic
Purity Grade ACS Grade
Linear Formula K2HPO4
Formula Weight 174.18
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC200901

 
lab chem
LC200901
500g Each for $172.63
Description & Specifications

Specifications

Assay ≥98.0%
Formula Weight 174.18
CAS Max % 100
CAS 7758-11-4
Name Note Dibasic
Physical Form Granules
Linear Formula K2HPO4
Health Hazard 2 GHS H Statement
Product is not hazardous.
Solubility Soluble in water
Melting Point 340°C
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Quantity 500g
Chemical Name or Material Potassium Phosphate
Purity Grade ACS Grade
Identification Passes Test
Color White
Packaging Poly Bottle
Storage Information Room Temperature
Decomposition Information Phosphorus oxides
EINECS Number 231-834-5
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.