Learn More
Potassium phosphate, dibasic, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Potassium phosphate, dibasic, 99+%, for analysis, anhydrous, ACROS Organics, Quantity: 5kg, Packaging: Plastic drum, CAS: 7758-11-4, White, Grade: Analysis, Melting Point: 340.0 deg.C, Molecular Formula: HK2O4P, Molecular Weight: 174.18, Percent Purity: 99+%, pH: 8.5 to 9.6 (5% soln. at 20 deg.C)
Supplier: Thermo Scientific Chemicals 215470010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysisSpecifications
Potassium phosphate, dibasic | |
7758-11-4 | |
8.5 to 9.6 (5% soln. at 20°C) | |
1 kg | |
HK2O4P | |
15,7779 | |
Solubility in water: 1600g/L (20°C). Other solubilities: slightly soluble in alcohol | |
OP(=O)([O-])[O-].[K+].[K+] | |
174.18 | |
CHEBI:32031 | |
≥99% | |
1% (130°C) max. |
340.0°C | |
White | |
Fine Crystalline Powder | |
99+% | |
K2HPO4 | |
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
dipotassium;hydrogen phosphate | |
24450 | |
174.18 | |
Analytical | |
Plastic bottle |
Chemical Identifiers
7758-11-4 | |
174.18 | |
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
CHEBI:32031 | |
OP(=O)([O-])[O-].[K+].[K+] |
HK2O4P | |
ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
24450 | |
dipotassium;hydrogen phosphate |
Safety and Handling
EINECSNumber : 231-834-5
RUO – Research Use Only