Please login to your online account to display your discounted pricing

Potassium Phosphate, Dibasic, 98.0% min., ACS, EMD Millipore

$194.43 - $1,751.23

Chemical Identifiers

Chemical Name or Material Potassium Phosphate, Dibasic
Name Note For ACS analysis
CAS 7758-11-4
Physical Form Granular Powder
Assay Percent Range 98.0% min.
Molecular Formula K2HPO4
Formula Weight 174.18
Synonym Potassium dihydrogen phosphate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MPX15701

 
emd millipore corporation
PX15701
500g Each for $194.43

MPX15707

 
emd millipore corporation
PX15707
12kg Each for $1,751.23
Description & Specifications

Specifications

Formula Weight 174.18
Physical Form Granular Powder
Color White
Quantity 500g
Name Note For ACS analysis
Assay Percent Range 98.0% min.
Chemical Name or Material Potassium Phosphate, Dibasic
CAS 7758-11-4
Synonym Potassium dihydrogen phosphate
Molecular Formula K2HPO4