missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Phosphate, Dibasic, 1.0M solution in water, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 397080025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Potassium phosphate, dibasic | |
2.5L | |
K2HPO4 | |
15, 7779 | |
ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
dipotassium;hydrogen phosphate | |
24450 | |
174.18 | |
1.13 |
7758-11-4,7732-18-5 | |
HK2O4P | |
MFCD00011383 | |
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
OP(=O)([O-])[O-].[K+].[K+] | |
174.18 | |
CHEBI:32031 | |
Glass bottle | |
1.1300g/mL |
RUO â Research Use Only