Please login to your online account to display your discounted pricing

Potassium Phosphate ACS AR Crystal, Macron Fine Chemicals™


macron

Quantity 125g
Packaging POLYSTORMOR

Manufacturer: avantor performance  710002

 View more versions of this product

Catalog No. MK7100125

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging POLYSTORMOR
Quantity 125g
Chemical Name or Material Potassium Phosphate
Name Note Monobasic
CAS 7778-77-0
Purity Grade ACS
Physical Form Crystalline
Molecular Formula KH2PO4
Formula Weight 136.09g/mol

Specifications

Chemical Name or Material Potassium Phosphate
Name Note Monobasic
CAS 7778-77-0
Purity Grade ACS
Physical Form Crystalline
Molecular Formula KH2PO4
Formula Weight 136.09g/mol