missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium phosphate, ACS, 98.0% min

Manufacturer:  Alfa Aesar™ 46644A7

 View more versions of this product

Catalog No. AA46644A7



Specifications

Specifications

Potassium phosphate
ACS Grade
K3O4P
14,7660
Soluble in water; Insoluble in alcohol;
[O-]P(=O)([O-])[O-].[K+].[K+].[K+]
212.265
212.27
2.564g/mL
White
5kg
7778-53-2
≥98.0%
MFCD00036295
Potassium phosphate, tribasic; Tripotassium phosphate
LWIHDJKSTIGBAC-UHFFFAOYSA-K
tripotassium;phosphate
62657
Granular
Hygroscopic
1340°C

Chemical Identifiers

7778-53-2
212.265
LWIHDJKSTIGBAC-UHFFFAOYSA-K
62657
[O-]P(=O)([O-])[O-].[K+].[K+].[K+]
K3O4P
MFCD00036295
Potassium phosphate, tribasic; Tripotassium phosphate
tripotassium;phosphate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : TC8450000

TSCA : TSCA

SDS Documents