missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium phosphate, 0.5M buffer soln., pH 7.6

$63.54 - $99.32

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.084
MDL Number MFCD00011401
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium;dihydrogen phosphate
SMILES OP(=O)(O)[O-].[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63189AK View Documents Alfa Aesar™
J63189AK
250mL Each for $63.54
 
AAJ63189AP View Documents Alfa Aesar™
J63189AP
500mL Each for $99.32
 
Description

Description

Applications
Potassium phosphate buffer is typically used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline. In addition to helping maintain pH, it supplies essential phosphate.

Solubility
Soluble in water, Insoluble in ethanol, alcohol.

Notes
Store sealed bottle containing the dehydrated medium at 2 - 30C. Once opened and recapped, place container in a low humidity environment at the same storage temperature. Protect from moisture and light by keeping container tightly closed.
Specifications

Chemical Identifiers

7778-77-0
136.084
GNSKLFRGEWLPPA-UHFFFAOYSA-M
CHEBI:63036
OP(=O)(O)[O-].[K+]
H2KO4P
MFCD00011401
516951
potassium;dihydrogen phosphate

Specifications

7778-77-0
MFCD00011401
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium;dihydrogen phosphate
516951
Liquid
250mL
H2KO4P
0.5M buffer soln.
OP(=O)(O)[O-].[K+]
136.084
CHEBI:63036
7.6
Potassium phosphate buffer soln.
Safety and Handling

Safety and Handling

TSCA : TSCA