Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium phosphate, 0.2M buffer soln., pH 7.4, low endotoxin

Chemical Identifiers

Chemical Name or Material Potassium phosphate
CAS 7778-77-0
Name Note low endotoxin
MDL Number MFCD00011401
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67035AE

 
alfa aesar™
J67035AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67035AK

 
alfa aesar™
J67035AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Potassium phosphate
Physical Form Liquid
MDL Number MFCD00011401
CAS 7778-77-0
pH 7.4
Color Colorless
Quantity 100mL
TSCA TSCA
Name Note low endotoxin
Concentration 0.2M buffer soln.