Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium phosphate, 0.2M buffer soln., pH 7.2, low endotoxin

Chemical Identifiers

Chemical Name or Material Potassium phosphate
CAS 7778-77-0
Name Note low endotoxin
MDL Number MFCD00011401
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67316AE

 
alfa aesar™
J67316AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67316AK

 
alfa aesar™
J67316AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Potassium phosphate
MDL Number MFCD00011401
Physical Form Liquid
Name Note low endotoxin
Concentration 0.2M buffer soln.
pH 7.2
Quantity 100mL
TSCA TSCA
Color Colorless
CAS 7778-77-0