Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
missing translation for 'pleaseLogIn'

Potassium Phenyltrifluoroborate, TCI America™

Manufacturer:  TCI America P158225G

 View more versions of this product

Catalog No. P158225G



Specifications

Specifications

Potassium Phenyltrifluoroborate
≥98.0% (HPLC,T)
MFCD01318172
[B-](C1=CC=CC=C1)(F)(F)F.[K+]
184.01
184.01
White
25g
153766-81-5
C6H5BF3K
DVAFPKUGAUFBTJ-UHFFFAOYSA-N
potassium;trifluoro(phenyl)boranuide
23675248
Crystal-Powder at 20°C
301°C

Chemical Identifiers

153766-81-5
184.01
DVAFPKUGAUFBTJ-UHFFFAOYSA-N
potassium;trifluoro(phenyl)boranuide
C6H5BF3K
MFCD01318172
23675248
[B-](C1=CC=CC=C1)(F)(F)F.[K+]
Safety and Handling

Safety and Handling

TSCA : No

Documents