Learn More
Potassium phenyltrifluoroborate, 98%, Thermo Scientific Chemicals
$62.64 - $170.22
Chemical Identifiers
CAS | 153766-81-5 |
---|---|
Molecular Formula | C6H5BF3K |
Molecular Weight (g/mol) | 184.01 |
MDL Number | MFCD01318172 |
InChI Key | DVAFPKUGAUFBTJ-UHFFFAOYSA-N |
Synonym | potassium phenyltrifluoroborate, potassium trifluoro phenyl borate, potassium trifluoro phenyl boranuide, pubchem11425, potassiumphenyltrifluoroborate, amtb100, phenyltrifluoropotassioboron v, potassium trifluoro phenyl borate 1-, potassium ion trifluoro phenyl boranuide, potassium tris fluoranyl-phenyl-boranuide |
PubChem CID | 23675248 |
IUPAC Name | potassium;trifluoro(phenyl)boranuide |
SMILES | [B-](C1=CC=CC=C1)(F)(F)F.[K+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1756803
|
Thermo Scientific Chemicals
L1756803 |
1 g |
Each for $62.64
|
|
|||||
AAL1756806
|
Thermo Scientific Chemicals
L1756806 |
5 g |
Each for $170.22
|
|
|||||
Description
Aryltrifluoroborate salts are more nucleophilic than the corresponding arylboronic acids and undergo ligand-free Pd-catalyzed cross-coupling reactions with arenediazonium tetrafluoroborates to give good yields of biaryls. Suzuki Cross-Coupling are conducted using organotrifluoroborates as a potent boronic acid surrogates. In the presence of DicarbonylÂ(2,4-pentanedionato)Ârhodium(I)Â, 39295, K aryl- and alkenyltrifluoroborates add to aldehydes and enones to give secondary alcohols and saturated ketones, respectively.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAryltrifluoroborate salts are more nucleophilic than the corresponding arylboronic acids and undergo ligand-free Pd-catalyzed cross-coupling reactions with arenediazonium tetrafluoroborates to give good yields of biaryls. Suzuki Cross-Coupling are conducted using organotrifluoroborates as a potent boronic acid surrogates. In the presence of DicarbonylÂ(2,4-pentanedionato)Ârhodium(I)Â, 39295, K aryl- and alkenyltrifluoroborates add to aldehydes and enones to give secondary alcohols and saturated ketones, respectively.
Solubility
Soluble in water.
Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Chemical Identifiers
153766-81-5 | |
184.01 | |
DVAFPKUGAUFBTJ-UHFFFAOYSA-N | |
23675248 | |
[B-](C1=CC=CC=C1)(F)(F)F.[K+] |
C6H5BF3K | |
MFCD01318172 | |
potassium phenyltrifluoroborate, potassium trifluoro phenyl borate, potassium trifluoro phenyl boranuide, pubchem11425, potassiumphenyltrifluoroborate, amtb100, phenyltrifluoropotassioboron v, potassium trifluoro phenyl borate 1-, potassium ion trifluoro phenyl boranuide, potassium tris fluoranyl-phenyl-boranuide | |
potassium;trifluoro(phenyl)boranuide |
Specifications
153766-81-5 | |
C6H5BF3K | |
1 g | |
potassium phenyltrifluoroborate, potassium trifluoro phenyl borate, potassium trifluoro phenyl boranuide, pubchem11425, potassiumphenyltrifluoroborate, amtb100, phenyltrifluoropotassioboron v, potassium trifluoro phenyl borate 1-, potassium ion trifluoro phenyl boranuide, potassium tris fluoranyl-phenyl-boranuide | |
DVAFPKUGAUFBTJ-UHFFFAOYSA-N | |
potassium;trifluoro(phenyl)boranuide | |
23675248 | |
98% |
296°C to 301°C | |
MFCD01318172 | |
7782070 | |
Soluble in water. | |
[B-](C1=CC=CC=C1)(F)(F)F.[K+] | |
184.01 | |
184 | |
Potassium phenyltrifluoroborate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only