Please login to your online account to display your discounted pricing

Potassium phenyltrifluoroborate, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Potassium phenyltrifluoroborate
CAS 153766-81-5
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C6H5BF3K
Formula Weight 184.01g/mol
MDL Number MFCD01318172
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC382290050

  SDS
fisher scientific acros
382290050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Quantity 5g
Chemical Name or Material Potassium phenyltrifluoroborate
CAS 153766-81-5
Physical Form White powder
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C6H5BF3K
Formula Weight 184.01g/mol
MDL Number MFCD01318172
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.