Please login to your online account to display your discounted pricing

Potassium Persulfate, BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

$329.60 - $951.47

Chemical Identifiers

Chemical Name or Material Potassium Persulfate
CAS 7727-21-1
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Molecular Formula K2S2O8
Formula Weight 270.32
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003796

 
avantor performance
323901
Wide Mouth Amber Glass Bottle 500g Each for $329.60

02003797

 
avantor performance
323905
Poly Bottle 2.5kg Each for $951.47
Description & Specifications

Specifications

Formula Weight 270.32
Chemical Name or Material Potassium Persulfate
Molecular Formula K2S2O8
CAS 7727-21-1
Packaging Wide Mouth Amber Glass Bottle
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Specific Gravity 1L = 2.48kg
Quantity 500g

    Suitable for Phosphate and Mercury Determinations