Please login to your online account to display your discounted pricing

Potassium Persulfate, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$181.34 - $706.47

Chemical Identifiers

Chemical Name or Material Potassium Persulfate
CAS 7727-21-1
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula K2S2O8
Formula Weight 270.32
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003795

 
avantor performance
323805
Poly Bottle 2.5kg Each for $706.47

02003794

 
avantor performance
323801
Wide Mouth Amber Glass Bottle 500g Each for $181.34
Description & Specifications

Specifications

Formula Weight 270.32
Chemical Name or Material Potassium Persulfate
Molecular Formula K2S2O8
CAS 7727-21-1
Packaging Poly Bottle
Purity Grade BAKER ANALYZED™ Reagent
Specific Gravity 1L = 2.48kg
Quantity 2.5kg