Please login to your online account to display your discounted pricing

Potassium Persulfate, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: avantor performance  323805

 View more versions of this product

Catalog No. 02003795

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 270.32
Chemical Name or Material Potassium Persulfate
CAS 7727-21-1
Molecular Formula K2S2O8
Purity Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.48kg
Quantity 2.5kg

Specifications

Chemical Name or Material Potassium Persulfate
CAS 7727-21-1
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula K2S2O8
Formula Weight 270.32