Please login to your online account to display your discounted pricing

Potassium Persulfate, ACS Grade, ≥ 99.0 %, LabChem™

$229.54 - $229.54

Chemical Identifiers

Chemical Name or Material Potassium Persulfate
CAS 7727-21-1
CAS Max % 100
Purity Grade ACS Grade
Linear Formula K2S2O8
Formula Weight 270.32
UN Number UN1492
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC200701

 
lab chem
LC200701
500g Each for $229.54
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 270.32
Chemical Name or Material Potassium Persulfate
EINECS Number 231-781-8
Identification Passes Test
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Crystals
Solubility Soluble in water
Specific Gravity 2.58g/cm3
Storage Information Room Temperature
Linear Formula K2S2O8
CAS 7727-21-1
UN Number UN1492
Decomposition Information Oxygen; Sulfur compounds
Color White
Melting Point 100°C
Quantity 500g
Health Hazard 1 Danger
CAS Max % 100
Health Hazard 2 GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Health Hazard 3 GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Avoid breathing dust.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Contaminated work clothing must not be allowed out of the workplace.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
If experiencing respiratory symptoms: Call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Density 2.58g/cm3
Safety and Handling

Danger

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Avoid breathing dust.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Contaminated work clothing must not be allowed out of the workplace.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
If experiencing respiratory symptoms: Call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.