Please login to your online account to display your discounted pricing

Potassium Persulfate ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Potassium Persulfate
CAS 7727-21-1
Purity Grade ACS Grade
Molecular Formula K2S2O8
Formula Weight 270.32
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK7076500

 
avantor performance
707612
Glass 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 270.32
Chemical Name or Material Potassium Persulfate
Packaging Glass
Molecular Formula K2S2O8
CAS 7727-21-1
Quantity 500g