Learn More
Potassium persulfate, 99+%, ACS reagent, Thermo Scientific Chemicals
$159.69 - $622.13
Chemical Identifiers
CAS | 7727-21-1 |
---|---|
Molecular Formula | K2O8S2 |
Molecular Weight (g/mol) | 270.3 |
MDL Number | MFCD00011386 |
InChI Key | USHAGKDGDHPEEY-UHFFFAOYSA-L |
Synonym | potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc |
PubChem CID | 24412 |
IUPAC Name | dipotassium;sulfonatooxy sulfate |
SMILES | [O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC424185000
|
Thermo Scientific Chemicals
424185000 |
500 g | Plastic bottle |
N/A
|
|
||||
AC424180025
|
Thermo Scientific Chemicals
424180025 |
2.5 kg | Plastic bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
7727-21-1 | |
270.3 | |
USHAGKDGDHPEEY-UHFFFAOYSA-L | |
24412 | |
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+] |
K2O8S2 | |
MFCD00011386 | |
potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc | |
dipotassium;sulfonatooxy sulfate |
Specifications
100.0°C | |
White | |
500 g | |
K2O8S2 | |
MFCD00011386 | |
15, 7777 | |
Solubility in water: 5g/100mL (20°C). Other solubilities: insoluble in alcohol | |
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+] | |
270.3 | |
270.3 | |
ACS Reagent | |
2.47 |
7727-21-1 | |
Fine Hygroscopic Crystalline Powder | |
99+% | |
K2S2O8 | |
08,417; 15,274 | |
potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc | |
USHAGKDGDHPEEY-UHFFFAOYSA-L | |
dipotassium;sulfonatooxy sulfate | |
24412 | |
≥99% | |
Plastic bottle | |
Potassium persulfate, 99+% |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficu
GHS P Statement
Keep/Store away from clothing/ combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,r
GHS Signal Word: Danger
EINECSNumber : 231-781-8
RTECSNumber : SE0400000
TSCA : TSCA
RUO – Research Use Only