Please login to your online account to display your discounted pricing

Potassium Persulfate, 5% (w/v) Aqueous Solution, APHA/ASTM/EPA for Mercury, Ricca Chemical

$134.12 - $134.12

Chemical Identifiers

Chemical Name or Material Potassium Persulfate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

6450-32

 
ricca chemical company
RI645032
Glass 1L Each for $134.12

6450-16

 
ricca chemical company
RI645016
Glass 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1L
Methods APHA/ASTM/EPA for Mercury
Chemical Name or Material Potassium Persulfate
Concentration or Composition (by Analyte or Components) 5% (w/v) Aqueous Solution
Packaging Glass