missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium perrhenate, 99% (metals basis), Re 64%

A powerful oxidizer

Manufacturer:  Alfa Aesar™ 1141118

 View more versions of this product

Catalog No. AA1141118



Specifications

Specifications

Potassium perrhenate
(metals basis)
MFCD00011365
acmc-1bx96
[O-][Re](=O)(=O)=O.[K+]
289.301
289.3
99%
4.887g/mL
1.643
White
50g
10466-65-6
KO4Re
UN1479
QFKRWIFGDGKWLM-UHFFFAOYSA-N
potassium;oxido(trioxo)rhenium
3661560
Crystalline
Re 64%
Odorless
1360°C to 1370°C
550°C

Chemical Identifiers

10466-65-6
289.301
QFKRWIFGDGKWLM-UHFFFAOYSA-N
3661560
[O-][Re](=O)(=O)=O.[K+]
KO4Re
MFCD00011365
acmc-1bx96
potassium;oxido(trioxo)rhenium
Safety and Handling

Safety and Handling

GHS H Statement
H272
May intensify fire
oxidizer.

GHS P Statement
P221-P210-P220-P280-P370+P378a-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

RTECSNumber : SE1400000

TSCA : TSCA

Documents