missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Peroxymonosulfate [> ca. 45%(T) as KHSO5], TCI America™

Manufacturer:  TCI America O031025G

 View more versions of this product

Catalog No. O031025G



Specifications

Specifications

Potassium Peroxymonosulfate [> ca. 45%(T) as KHSO5]
H3K5O18S4
Oxone
Crystal-Powder at 20°C
2.3 (10g/L H2O soln. 77°C)
37222-66-5
3260
614.74
White
25g

Chemical Identifiers

37222-66-5
H3K5O18S4
Oxone
Safety and Handling

Safety and Handling

TSCA : No

Documents