missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium peroxydisulfate, ACS, 99.0% min, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04693930

 View more versions of this product

Catalog No. AA4693930



Description

Description

Potassium persulfate is a powerful oxidant, commonly used to initiate polymerizations. Potassium Persulphate used for bleaching and textile desizing, as an oxidizing agent and antiseptic, in purification of ammonium sulfate, and in the manufacture of soap and pharmaceuticals. It is also used as al laboratory oxidant and photography chemical. It is a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium peroxydisulfate
Low nitrogen
White
250 g
K2S2O8
UN1492
potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc
USHAGKDGDHPEEY-UHFFFAOYSA-L
dipotassium;sulfonatooxy sulfate
24412
≥99.0%
Odorless
100°C (dec.)
7727-21-1
Crystalline
K2O8S2
MFCD00011386
14,7656
It is highly soluble in water. It is insoluble in alcohol.
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+]
270.309
270.31
ACS
2.477 g/mL

Chemical Identifiers

7727-21-1
270.309
USHAGKDGDHPEEY-UHFFFAOYSA-L
24412
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+]
K2O8S2
MFCD00011386
potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc
dipotassium;sulfonatooxy sulfate
Safety and Handling

Safety and Handling

GHS H Statement
H334-H272-H302-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May intensify fire
oxidizer.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P210-P220-P221-P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P370+P378q-P501c

H272-H302-H315-H317-H319-H334-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: POTASSIUM PERSULFATE

EINECSNumber : 231-781-8

RTECSNumber : SE0400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only