Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium peroxydisulfate, Electrophoresis Grade, 99.0%


Quantity 5g

Manufacturer: alfa aesar™  J6667206

 View more versions of this product

Catalog No. AAJ6667206

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 270.31
Density 2.477g/mL
Assay 99.0%
CAS 7727-21-1
Health Hazard 3 GHS P Statement
P221-P210-P285-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
In case of inadequate ventilation wear respiratory protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Odorless
MDL Number MFCD00011386
Color White
Quantity 5g
Health Hazard 1 Danger
RTECS Number SE0400000
TSCA TSCA
Purity Grade Electrophoresis Grade
Physical Form Crystalline
UN Number UN1492
Merck Index 14,7656
Health Hazard 2 GHS H Statement
H334-H272-H302-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May intensify fire
oxidizer.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Chemical Name or Material Potassium peroxydisulfate
Melting Point 100°C (dec.)
DOT Information Transport Hazard Class: 5.1; Packaging Group: III; Proper Shipping Name: POTASSIUM PERSULFATE

Specifications

Chemical Name or Material Potassium peroxydisulfate
CAS 7727-21-1
Purity Grade Electrophoresis Grade
Formula Weight 270.31
MDL Number MFCD00011386
UN Number UN1492
Merck Index 14,7656
Accessories

Danger

GHS P Statement
P221-P210-P285-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
In case of inadequate ventilation wear respiratory protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H272-H302-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May intensify fire
oxidizer.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.