Learn More
Potassium peroxydisulfate, Electrophoresis Grade, 99.0%, Thermo Scientific Chemicals
$57.81 - $73.25
Chemical Identifiers
CAS | 7727-21-1 |
---|---|
Molecular Formula | K2O8S2 |
Molecular Weight (g/mol) | 270.309 |
MDL Number | MFCD00011386 |
InChI Key | USHAGKDGDHPEEY-UHFFFAOYSA-L |
Synonym | potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc |
PubChem CID | 24412 |
IUPAC Name | dipotassium;sulfonatooxy sulfate |
SMILES | [O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6667214
|
Thermo Scientific Chemicals
J6667214 |
25 g |
Each for $57.81
|
|
|||||
AAJ6667222
|
Thermo Scientific Chemicals
J6667222 |
100 g |
Each for $73.25
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7727-21-1 | |
270.309 | |
USHAGKDGDHPEEY-UHFFFAOYSA-L | |
24412 | |
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+] |
K2O8S2 | |
MFCD00011386 | |
potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc | |
dipotassium;sulfonatooxy sulfate |
Specifications
100°C (decomposition) | |
White | |
25 g | |
MFCD00011386 | |
14,7656 | |
USHAGKDGDHPEEY-UHFFFAOYSA-L | |
dipotassium;sulfonatooxy sulfate | |
24412 | |
99.0% | |
Odorless | |
Potassium peroxydisulfate |
7727-21-1 | |
Crystalline | |
K2O8S2 | |
UN1492 | |
potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc | |
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+] | |
270.309 | |
270.31 | |
Electrophoresis | |
2.477 g/mL |
Safety and Handling
GHS H Statement
H334-H272-H302-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May intensify fire
oxidizer.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P210-P220-P221-P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P370+P378q-P501c
H272-H302-H315-H317-H319-H334-H335
DOTInformation : Transport Hazard Class: 5.1; Packaging Group: III; Proper Shipping Name: POTASSIUM PERSULFATE
EINECSNumber : 231-781-8
RTECSNumber : SE0400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only