missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium peroxydisulfate, ACS, 99.0% min., Alfa Aesar™
A Polymerization Accelerant For PAGE Gels.
Manufacturer: Alfa Aesar 1314530
Specifications
Potassium peroxydisulfate | |
99.0% min. | |
MFCD00011386 | |
potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc | |
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+] | |
270.309 | |
270.31 | |
ACS | |
Maximum level of impurities: Insoluble matter 0.005%, Chlorine compounds (as Cl) 0.001%, Heavy Metals (as Pb) 0.001%, Fe 5ppm, Mn 2ppm. | |
White |
7727-21-1 | |
K2O8S2 | |
White crystalline powder | |
USHAGKDGDHPEEY-UHFFFAOYSA-L | |
dipotassium;sulfonatooxy sulfate | |
24412 | |
Crystalline Powder | |
2.477g/cm3 | |
Odorless | |
250g |
Chemical Identifiers
7727-21-1 | |
270.309 | |
USHAGKDGDHPEEY-UHFFFAOYSA-L | |
24412 | |
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+] |
K2O8S2 | |
MFCD00011386 | |
potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc | |
dipotassium;sulfonatooxy sulfate |
Safety and Handling
Do not breathe dust. Avoid contact with skin. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
H334-H315-H319-H317-H272-H302-H335: May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May intensify fire; oxidizer. Harmful if swallowed. May cause respiratory irritation.
DOTInformation : Hazard Class: 5.1, Packing Group: III
EINECSNumber : 231-781-8