Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium peroxydisulfate, 99.99% (metals basis)

$155.84 - $155.84

Chemical Identifiers

Chemical Name or Material Potassium peroxydisulfate
CAS 7727-21-1
Formula Weight 270.31g/mol
MDL Number MFCD00011386
UN Number UN1492
Merck Index 14,7656
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1822722

 
alfa aesar™
18227-22
100g Each for $155.84
Description & Specifications

Specifications

Density 2.477g/mL
Odor Odorless
Quantity 100g
Melting Point 100°C (dec.)
Quantity 100g
Chemical Name or Material Potassium peroxydisulfate
CAS 7727-21-1
Physical Form Crystalline
Assay 99.99%
Assay Percent Notes (metals basis)
Formula Weight 270.31g/mol
MDL Number MFCD00011386
UN Number UN1492
Merck Index 14,7656
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H272-H302-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May intensify fire
oxidizer.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P221-P210-P284a-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: POTASSIUM PERSULFATE
RTECS Number SE0400000
TSCA TSCA