missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium periodate, 99%

Manufacturer:  Alfa Aesar™ A1130822

 View more versions of this product

Catalog No. AAA1130822


 

Description

Description

Applications
Potassium periodate is used as an oxidizing agent in organic synthesis and an analytical reagent for the determination of potassium and cerium. It is also used in colorimetric estimation of manganese. It reacts with ammonium cerium(IV) nitrate to prepare tris[trinitratocerium(IV)] paraperiodate which finds application as an oxidant.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong reducing agents, powdered metals and amines.
Specifications

Specifications

Potassium periodate
99%
MFCD00011407
14,7654
FJVZDOGVDJCCCR-UHFFFAOYSA-M
potassium;periodate
516896
582°C
7790-21-8
IKO4
UN1479
Potassium metaperiodate
[O-]I(=O)(=O)=O.[K+]
229.999
230
100g

Chemical Identifiers

7790-21-8
229.999
FJVZDOGVDJCCCR-UHFFFAOYSA-M
516896
IKO4
MFCD00011407
Potassium metaperiodate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

TSCA : TSCA

SDS Documents