missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium periodate, 99.8%-100.3%, ACS, Alfa Aesar™

Manufacturer:  Alfa Aesar 2289614

 View more versions of this product

Catalog No. AA2289614



Specifications

Specifications

Potassium periodate
7790-21-8
IKO4
potassium periodate, potassium metaperiodate, unii-q3loc6p66l, q3loc6p66l, periodic acid hio4 , potassium salt, potassium ion io4, ssmicroaea 1/4 o, acmc-20ajue, ssmicroaea 1/4 oni, periodic acid hio4 , potassium salt 1:1
[O-]I(=O)(=O)=O.[K+]
229.999
230
25g
For ACS analysis
99.8%-100.3%
MFCD00011407
FJVZDOGVDJCCCR-UHFFFAOYSA-M
potassium;periodate
516896
Powder

Chemical Identifiers

7790-21-8
229.999
FJVZDOGVDJCCCR-UHFFFAOYSA-M
516896
[O-]I(=O)(=O)=O.[K+]
IKO4
MFCD00011407
potassium periodate, potassium metaperiodate, unii-q3loc6p66l, q3loc6p66l, periodic acid hio4 , potassium salt, potassium ion io4, ssmicroaea 1/4 o, acmc-20ajue, ssmicroaea 1/4 oni, periodic acid hio4 , potassium salt 1:1
potassium;periodate
Safety and Handling

Safety and Handling

EINECSNumber : 232-196-0

SDS Documents