missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium perchlorate, anhydrous, ACS, 99.0 to 100.5%, Alfa Aesar™

For Parr Bomb Work

Manufacturer:  Thermo Fisher Scientific 1163022

 View more versions of this product

Catalog No. AA1163022

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Potassium perchlorate, anhydrous
7778-74-7
ClKO4
White powder
Partially soluble in water. Insoluble in alcohol.Soluble in water. Slightly soluble in acetone. Insoluble in ethanol,ethyl acetate and ether.
[O-]Cl(=O)(=O)=O.[K+]
138.544
138.55
ACS
Odorless
Black
100g
For analysis ACS
99.0-100.5%
MFCD00011362
astrumal, irenat, perchloracap, perchloric acid, potassium salt, perchloric acid, potassium salt 1:1, peroidin, potassium hyperchloride, potassium perchlorate, potassium perchlorate kclo4, potassium perchlorate usp
YLMGFJXSLBMXHK-UHFFFAOYSA-M
potassium;perchlorate
516900
-100 Mesh Powder
2.52 g/cm3 at 20°C
Hygroscopic
400°C (decomposition)

Chemical Identifiers

7778-74-7
138.544
YLMGFJXSLBMXHK-UHFFFAOYSA-M
516900
[O-]Cl(=O)(=O)=O.[K+]
ClKO4
MFCD00011362
astrumal, irenat, perchloracap, perchloric acid, potassium salt, perchloric acid, potassium salt 1:1, peroidin, potassium hyperchloride, potassium perchlorate, potassium perchlorate kclo4, potassium perchlorate usp
potassium;perchlorate
Safety and Handling

Safety and Handling

P210-P220-P221-P264b-P270-P280-P283-P301+P312-P306+P360-P330-P370+P378q-P371+P380+P375-P501c

H271-H302

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 231-912-9

RTECSNumber : SC9700000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents