missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium perchlorate, anhydrous, ACS, 99.0-100.5%, Thermo Scientific Chemicals

For Parr Bomb Work

Supplier:  Thermo Scientific Chemicals 01163036

 View more versions of this product

Catalog No. AA1163036

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

For Parr Bomb WorkPotassium perchlorate is used in ammunition percussion caps, explosive primers, fireworks and propellants. It acts as a solid rocket propellant. It is also used as an oxidizer in colored pyrotechnic compositions and flash powder. It is used as an antithyroid and involved in the treatment of hyperthyroidism. The mixture of anthracene and sulfur with potassium perchlorate is used for generating black smoke signaling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium perchlorate, anhydrous
For analysis ACS
Black
500 g
ClKO4
potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp
YLMGFJXSLBMXHK-UHFFFAOYSA-M
potassium;perchlorate
516900
ACS
Hygroscopic
2.52
400°C (decomposition)
7778-74-7
Powder
99.0-100.5%
MFCD00011362
Partially soluble in water. Insoluble in alcohol.Soluble in water. Slightly soluble in acetone. Insoluble in ethanol,ethyl acetate and ether.
[K+].[O-][Cl](=O)(=O)=O
138.54
138.55
Odorless
White powder

Chemical Identifiers

7778-74-7
138.54
YLMGFJXSLBMXHK-UHFFFAOYSA-M
516900
[K+].[O-][Cl](=O)(=O)=O
ClKO4
MFCD00011362
potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp
potassium;perchlorate
Safety and Handling

Safety and Handling

P210-P220-P221-P264b-P270-P280-P283-P301+P312-P306+P360-P330-P370+P378q-P371+P380+P375-P501c

H271-H302

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 231-912-9

RTECSNumber : SC9700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only