missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ Potassium pentachlororuthenate(III) hydrate, 99.95% (metals basis), Ru 25.4% min
Manufacturer: Alfa Aesar™ 1217303
Specifications
Potassium pentachlororuthenate(III) hydrate | |
(metals basis) | |
MFCD00058747 | |
SRVGBBZEPHUDFZ-UHFFFAOYSA-I | |
dipotassium;pentachlororuthenium(2-);hydrate | |
16211344 | |
Crystalline | |
Ru 25.4% min. |
14404-33-2 | |
Cl5H2K2ORu | |
dipotassium aquapentachlororuthenate, potassium pentachlororuthenate iii hydrate, potassium pentachlororuthenate iii hydrate, dipotassium aquapentachlororuthenate iii, dipotassium pentachlororutheniumdiuide hydrate, potassium aquapentachlororuthenate iii , premion, ruthenate 2-, aquapentachloro-, dipotassium, oc-6-21 | |
O.Cl[Ru-2](Cl)(Cl)(Cl)Cl.[K+].[K+] | |
374.532 | |
356.54 (Anhydrous) | |
99.95% | |
1g |
Chemical Identifiers
14404-33-2 | |
374.532 | |
SRVGBBZEPHUDFZ-UHFFFAOYSA-I | |
16211344 | |
O.Cl[Ru-2](Cl)(Cl)(Cl)Cl.[K+].[K+] |
Cl5H2K2ORu | |
MFCD00058747 | |
dipotassium aquapentachlororuthenate, potassium pentachlororuthenate iii hydrate, potassium pentachlororuthenate iii hydrate, dipotassium aquapentachlororuthenate iii, dipotassium pentachlororutheniumdiuide hydrate, potassium aquapentachlororuthenate iii , premion, ruthenate 2-, aquapentachloro-, dipotassium, oc-6-21 | |
dipotassium;pentachlororuthenium(2-);hydrate |
Safety and Handling
TSCA : TSCA