missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium pentachlororuthenate(III) hydrate, 99.95% (metals basis), Ru 25.4% min

Manufacturer:  Alfa Aesar™ 1217306

 View more versions of this product

Catalog No. AA1217306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Potassium pentachlororuthenate(III) hydrate
(metals basis)
MFCD00058747
SRVGBBZEPHUDFZ-UHFFFAOYSA-I
dipotassium;pentachlororuthenium(2-);hydrate
16211344
Crystalline
Ru 25.4% min.
14404-33-2
Cl5H2K2ORu
dipotassium aquapentachlororuthenate, potassium pentachlororuthenate iii hydrate, potassium pentachlororuthenate iii hydrate, dipotassium aquapentachlororuthenate iii, dipotassium pentachlororutheniumdiuide hydrate, potassium aquapentachlororuthenate iii , premion, ruthenate 2-, aquapentachloro-, dipotassium, oc-6-21
O.Cl[Ru-2](Cl)(Cl)(Cl)Cl.[K+].[K+]
374.532
356.54 (Anhydrous)
99.95%
5g

Chemical Identifiers

14404-33-2
374.532
SRVGBBZEPHUDFZ-UHFFFAOYSA-I
16211344
O.Cl[Ru-2](Cl)(Cl)(Cl)Cl.[K+].[K+]
Cl5H2K2ORu
MFCD00058747
dipotassium aquapentachlororuthenate, potassium pentachlororuthenate iii hydrate, potassium pentachlororuthenate iii hydrate, dipotassium aquapentachlororuthenate iii, dipotassium pentachlororutheniumdiuide hydrate, potassium aquapentachlororuthenate iii , premion, ruthenate 2-, aquapentachloro-, dipotassium, oc-6-21
dipotassium;pentachlororuthenium(2-);hydrate
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents