Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium pentachlororhodate(III), Premion™, 99.99% (metals basis), Rh 28.2% min

Chemical Identifiers

Chemical Name or Material Potassium pentachlororhodate(III)
CAS 65980-75-8
Purity Grade Premion™
Formula Weight 358.37
MDL Number MFCD00049661
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4040903

 
alfa aesar™
4040903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4040906

 
alfa aesar™
4040906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 358.37
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P321-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see ... on this label).Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Potassium pentachlororhodate(III)
Physical Form Powder
Assay 99.99%
MDL Number MFCD00049661
CAS 65980-75-8
Quantity 1g
Health Hazard 2 GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Concentration Rh 28.2% min.
RTECS Number VI8580332
Purity Grade Premion™
Assay Percent Notes (metals basis)
Solubility Information Slightly soluble in cold water; Insoluble in alcohol
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P321-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see ... on this label).Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.