missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium pentachloronitrosylruthenate(II), Ru 25.8%

Manufacturer:  Alfa Aesar™ 1053401

 View more versions of this product

Catalog No. AA1053401



Specifications

Specifications

Potassium pentachloronitrosylruthenate(II)
Cl5K2NORu-
di-potassium nitrosylpentachlororuthenate ii, dipotassium pentachloro nitrosyl ruthenate ii, dipotassium pentachloro nitroso rutheniumdiuide, potassium pentachloronitrosylruthenate ii , ru
[N-]=O.Cl[Ru-2](Cl)(Cl)(Cl)Cl.[K+].[K+]
386.523
386.55
Ru 25.8%
0.25g
14854-54-7
MFCD00049789
CPBBXEQEBCUXGZ-UHFFFAOYSA-I
dipotassium;nitroxyl anion;pentachlororuthenium(2-)
71311320
Crystalline
220°C (decomposition)

Chemical Identifiers

14854-54-7
386.523
CPBBXEQEBCUXGZ-UHFFFAOYSA-I
71311320
[N-]=O.Cl[Ru-2](Cl)(Cl)(Cl)Cl.[K+].[K+]
Cl5K2NORu-
MFCD00049789
di-potassium nitrosylpentachlororuthenate ii, dipotassium pentachloro nitrosyl ruthenate ii, dipotassium pentachloro nitroso rutheniumdiuide, potassium pentachloronitrosylruthenate ii , ru
dipotassium;nitroxyl anion;pentachlororuthenium(2-)
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents