missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium pentaborate octahydrate, 97%

$52.50 - $52.50

Chemical Identifiers

CAS 12229-13-9
Formula Weight 586.43 (442.30 Anhydrous)
MDL Number MFCD00213879
Synonym Potassium borate
Chemical Name or Material Potassium pentaborate octahydrate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3943736 SDS Alfa Aesar™
3943736*
500g Each for $52.50
AA39437A3 SDS Alfa Aesar™
39437A3
2kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

12229-13-9
MFCD00213879
Potassium pentaborate octahydrate
586.43 (442.30 Anhydrous)
Potassium borate

Specifications

586.43 (442.30 Anhydrous)
Potassium borate
12229-13-9
500g
MFCD00213879
97%
780°C
Insoluble in water
White
Solid
Potassium pentaborate octahydrate

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA