missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium p-Tolyl Sulfate, TCI America™

Manufacturer:  TCI America P20915G

 View more versions of this product

Catalog No. P20915G



Specifications

Specifications

Potassium p-Tolyl Sulfate
≥98.0% (HPLC,T)
Potassium p-Cresyl Sulfate, Sulfuric Acid p-Tolyl Ester Potassium Salt, Sulfuric Acid p-Cresyl Ester Potassium Salt
CC1=CC=C(C=C1)OS(=O)(=O)[O-].[K+]
226.287
226.29
White
91978-69-7
C7H7KO4S
HTSFIPMTBJHYFQ-UHFFFAOYSA-M
potassium;(4-methylphenyl) sulfate
91972090
Crystal-Powder at 20°C
5g

Chemical Identifiers

91978-69-7
226.287
Potassium p-Cresyl Sulfate, Sulfuric Acid p-Tolyl Ester Potassium Salt, Sulfuric Acid p-Cresyl Ester Potassium Salt
potassium;(4-methylphenyl) sulfate
C7H7KO4S
HTSFIPMTBJHYFQ-UHFFFAOYSA-M
91972090
CC1=CC=C(C=C1)OS(=O)(=O)[O-].[K+]
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

Documents