Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium oxide, super, 96.5%

Chemical Identifiers

Chemical Name or Material Potassium oxide
CAS 12030-88-5
Name Note Super
Formula Weight 71.1
MDL Number MFCD00082411
UN Number UN2466
Synonym Potassium superoxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1405818

  SDS
alfa aesar™
1405818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1405830

  SDS
alfa aesar™
1405830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA14058A1

  SDS
alfa aesar™
14058A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 71.1
Chemical Name or Material Potassium oxide
Odor Odorless
Name Note Super
Sensitivity Air & Moisture Sensitive
MDL Number MFCD00082411
UN Number UN2466
Health Hazard 2 GHS H Statement
H271-H314-H318
May cause fire or explosion
strong oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Density 2.14g/mL
TSCA TSCA
Particle Size 2 to 10 Micron
Physical Form Powder
Assay 96.5%
CAS 12030-88-5
Synonym Potassium superoxide
Melting Point ≈400°C
Quantity 50g
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P221-P283-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Wear fire/flame resistant/retardant clothing.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 5.1; Packing Group: I; Proper Shipping Name: POTASSIUM SUPEROXIDE
Safety and Handling

Danger

GHS P Statement
P221-P283-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Wear fire/flame resistant/retardant clothing.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H271-H314-H318
May cause fire or explosion
strong oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.