missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium oxalate monohydrate, 99%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 424020025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium oxalate monohydrate | |
356.0°C | |
Authentic | |
Plastic bottle | |
KO2CCO2K·H2O | |
2.5 kg | |
potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate | |
QCPTVXCMROGZOL-UHFFFAOYSA-L | |
dipotassium;oxalate;hydrate | |
2724193 | |
99% | |
0.01% max |
6487-48-5 | |
White | |
99% | |
C2H2K2O5 | |
MFCD00150033 | |
15, 7772 | |
Solubility in water: 364g/L (20°C) | |
O.[K+].[K+].[O-]C(=O)C([O-])=O | |
184.23 | |
184.24 | |
ACS Reagent | |
Crystalline Powder |
Chemical Identifiers
6487-48-5 | |
184.23 | |
QCPTVXCMROGZOL-UHFFFAOYSA-L | |
2724193 | |
O.[K+].[K+].[O-]C(=O)C([O-])=O |
C2H2K2O5 | |
MFCD00150033 | |
potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate | |
dipotassium;oxalate;hydrate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Warning
RUO – Research Use Only