missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Oxalate Monohydrate (Crystalline/Certified ACS), Fisher Chemical

$254.77 - $274.85

Chemical Identifiers

CAS 6487-48-5
Physical Form Solid
Assay Percent Range 98.5 to 101.0%
Molecular Formula C2K2O4 · H2O
Formula Weight 184.23
MDL Number MFCD00150033
Synonym Oxalic Acid Potassium Salt
Chemical Name or Material Potassium Oxalate Monohydrate
Assay 98.5 to 101.0%
Grade Certified ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
P273-250 SDS Fisher Scientific Company
P273250
Glass Bottle 250g Each for $254.77
Case of 6 Each for $1,528.62
P273-500 SDS Fisher Scientific Company
P273500
Glass Bottle 500g Each for $274.85
Case of 6 Each for $1,649.10
Specifications Description & Specifications

Chemical Identifiers

6487-48-5
98.5 to 101.0%
184.23
Oxalic Acid Potassium Salt
98.5 to 101.0%
Solid
C2K2O4 · H2O
MFCD00150033
Potassium Oxalate Monohydrate
Certified ACS Grade

Specifications

6487-48-5
0.002% max.
Solid
98.5 to 101.0%
C2K2O4 · H2O
184.23
2811
Oxalic Acid Potassium Salt
0.02% max.
0.01% max.
White
7.0 to 8.5
Potassium Oxalate Monohydrate
Certified ACS Grade
0.002% max.
0.002% max.
Pass Test
0.01% max.
0.001% max.
MFCD00150033
Pass Test
Glass Bottle
Pass Test
2.13g/cm³
356°C
250g
98.5 to 101.0%

Safety and Handling

DOTInformation : DOT Class 6.1, : Poison