missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Oxalate Monohydrate, ACS Reagent, Reagents
Potassium Oxalate Monohydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C1287500500C2
Specifications
| 365°C | |
| Colorless/White | |
| Solid | |
| C2H2K2O5 | |
| Dipotassium Ethanedioic Acid | |
| O.[K+].[K+].[O-]C(=O)C([O-])=O | |
| 184.23 | |
| Glass Bottle |
| Potassium Oxalate Monohydrate, ACS Reagent | |
| 6487-48-5 | |
| 500 g | |
| MFCD00150033 | |
| QCPTVXCMROGZOL-UHFFFAOYSA-L | |
| dipotassium hydrate oxalate | |
| ACS | |
| 2.13 |
Chemical Identifiers
| 6487-48-5 | |
| 184.23 | |
| QCPTVXCMROGZOL-UHFFFAOYSA-L | |
| dipotassium hydrate oxalate |
| C2H2K2O5 | |
| MFCD00150033 | |
| Dipotassium Ethanedioic Acid | |
| O.[K+].[K+].[O-]C(=O)C([O-])=O |