missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Oxalate, Monohydrate, ACS Grade, 98.5% to 101.0%, LabChem™

$163.03 - $163.03

Chemical Identifiers

CAS 6487-48-5
CAS Max % 100
Linear Formula K2C2O4·H2O
Formula Weight 184.24
Chemical Name or Material Potassium Oxalate, Monohydrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC198351 Lab Chem
LC198351
500g Each for $163.03
Specifications Description & Specifications

Chemical Identifiers

6487-48-5
K2C2O4·H2O
Potassium Oxalate, Monohydrate
100
184.24
ACS Grade

Specifications

184.24
6487-48-5
2130kg/m3
Poly Bottle
White
500g
K2C2O4·H2O
98.5 to 101.0%
Soluble in water
Crystals
2,130kg/m3
Room Temperature
Passes Test
100
Potassium Oxalate, Monohydrate
ACS Grade

Safety and Handling

GHS H Statement
Harmful if swallowed or in contact with skin.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Call a poison center or doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off contaminated clothing and wash before reuse.
Dispose of contents/container to comply with local, state and federal regulations.

Warning

EINECSNumber : 209-506-8