missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium oxalate monohydrate, ACS, 98.8-101.0%

Manufacturer:  Alfa Aesar™ 1345236

 View more versions of this product

Catalog No. AA1345236



Specifications

Specifications

6 to 8.5
2.13g/mL
White
500g
C2H2K2O5
MFCD00150033
14,7651
Soluble in water
C(=O)(C(=O)[O-])[O-].O.[K+].[K+]
184.23
184.23 (166.22 Anhydrous)
98.8 to 101.0%
Potassium oxalate monohydrate
Odorless
>300°C
6487-48-5
K2C2O4·H2O
3752576
potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
QCPTVXCMROGZOL-UHFFFAOYSA-L
dipotassium;oxalate;hydrate
2724193
Granular/Powder
ACS

Chemical Identifiers

6487-48-5
184.23
QCPTVXCMROGZOL-UHFFFAOYSA-L
2724193
C(=O)(C(=O)[O-])[O-].O.[K+].[K+]
C2H2K2O5
MFCD00150033
potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
dipotassium;oxalate;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : RO2885000

TSCA : TSCA

Documents