Please login to your online account to display your discounted pricing

Potassium oxalate monohydrate, 99+%, for analysis, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC207710025

 View more versions of this product

Catalog No. AC207710025

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99%
Purity Grade For analysis
Physical Form Crystals or Flakes
Formula Weight 184.24
Quantity 2.5kg
Color White
pH 7 to 8.5 (5% soln. at 20°C)
Merck Index 15, 7772
Packaging Plastic bottle
Heavy Metals (as Pb) 20ppm max.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Health Hazard 1 GHS Signal Word: Warning
Sulfate 100ppm max.
Synonym Oxalic acid, dipotassium salt monohydrate
Iron (Fe) 10ppm max.
Infrared Spectrum Authentic
Chlorine (Cl) 50ppm max.
Assay Percent Range 99+%
Chemical Name or Material Potassium oxalate monohydrate
Molecular Formula C2K2O4·H2O
Linear Formula KO2CCO2K·H2O
MDL Number MFCD00150033
Name Note p.a.
CAS 6487-48-5

Specifications

Chemical Name or Material Potassium oxalate monohydrate
Name Note p.a.
CAS 6487-48-5
Purity Grade For analysis
Assay ≥99%
Assay Percent Range 99+%
Molecular Formula C2K2O4·H2O
Linear Formula KO2CCO2K·H2O
Formula Weight 184.24
MDL Number MFCD00150033
Synonym Oxalic acid, dipotassium salt monohydrate
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.

SDS & Safety Documents